Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Propanediamine-N,N,N',N'-tetraacetic Acid 98.0+%, TCI America™
SDP

Catalog No. P1605500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1605500G 500 g
P160525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1605500G Supplier TCI America Supplier No. P1605500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1939-36-2 | C11H18N2O8 | 306.27 g/mol

Chemical Identifiers

CAS 1939-36-2
Molecular Formula C11H18N2O8
Molecular Weight (g/mol) 306.27
MDL Number MFCD00210039
InChI Key DMQQXDPCRUGSQB-UHFFFAOYSA-N
PubChem CID 80296
IUPAC Name 2-({3-[bis(carboxymethyl)amino]propyl}(carboxymethyl)amino)acetic acid
SMILES OC(=O)CN(CCCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

Melting Point 227°C
Color White
Quantity 500 g
Synonym Trimethylenediamine-N,N,N′,N′-tetraacetic Acid
Formula Weight 306.27
Percent Purity ≥98.0% (T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Propanediamine-N,N,N′,N′-tetraacetic Acid
EINECSNumber (2)-3632
RTECSNumber MC1082500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.