Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Propanedisulfonic Acid (50-60% in Water), TCI America™
SDP

Catalog No. P120410G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P120410G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P120410G Supplier TCI America Supplier No. P120410G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 21668-77-9
Molecular Formula C3H6O6S2
Molecular Weight (g/mol) 202.20
MDL Number MFCD00191482
InChI Key MGNVWUDMMXZUDI-UHFFFAOYSA-L
Synonym 1,3-propanedisulfonic acid, eprodisate, 1,3-propanedisulfonic acid in water, unii-6qfp76v7s7, propane-1,3-disulfonate, eprodisate inn, 1,3-propanesulfonic acid, 1,3-propanedisulfonicacid, 1,3-propanedisulphonic acid, 1,3 propanesulfonate
PubChem CID 428573
IUPAC Name propane-1,3-disulfonate
SMILES [O-]S(=O)(=O)CCCS([O-])(=O)=O

Specifications

Color Yellow
Quantity 10 g
UN Number 3265
Formula Weight 204.21
Physical Form Liquid
Chemical Name or Material 1,3-Propanedisulfonic Acid (50-60% in Water)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.