Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,3-Propanedisulfonic acid disodium salt

Catalog No. P857925G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P857925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P857925G Supplier Sigma Aldrich Supplier No. P857925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-phenyl-1-butanol undergoes biotransformation to aldehydes by oxidation in the presence of gluconobacteroxydans dsm 2343
  • It undergoes enantioselective transesterification reaction with vinyl acetate catalyzed by lipase isolated from pseudomonas cepacia

Specifications

CAS 36589-58-9
Molecular Formula C3H6Na2O6S2
Linear Formula NaSO3CH2CH2CH2SO3Na
MDL Number MFCD00191483
Quantity 5 g
Synonym Eprodisate disodium; NC 503
Molecular Weight (g/mol) 248.19
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.