Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,3-Propanesultone

Catalog No. 501844355
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-184-4355 5 g
50-289-9502 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-184-4355 Supplier Sigma Aldrich Supplier No. 2912505G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,3-Propanesultone
TRUSTED_SUSTAINABILITY

Specifications

CAS 1120-71-4
Melting Point 30°C to 33°C (lit.)
Density 1.392 g/mL (at 25°C (literature))
Boiling Point 180°C (30 mmHg)
Molecular Formula C3H6O3S
Linear Formula C3H6O3S
MDL Number MFCD00005355
Quantity 5 g
Synonym 3-Hydroxypropanesulfonic acid gamma-sultone
Molecular Weight (g/mol) 122.14
Percent Purity ≥99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.