Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,3-Propanesultone

Catalog No. 502914312
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-291-4312 1 kg
50-289-7284 25 g
50-289-7245 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-291-4312 Supplier Sigma Aldrich Supplier No. P507061KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,3-Propanesultone
TRUSTED_SUSTAINABILITY

Specifications

CAS 1120-71-4
Melting Point 30°C to 33°C (lit.)
Density 1.392 g/mL (at 25°C (literature))
Boiling Point 180°C (30 mmHg)
Molecular Formula C3H6O3S
Linear Formula C3H6O3S
MDL Number MFCD00005355
Quantity 1 kg
Synonym 3-Hydroxypropanesulfonic acid gamma-sultone
Molecular Weight (g/mol) 122.14
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.