Learn More
Cayman Chemical 1Octnoyl2Decnoyl3Stear 1mg
SDP
Supplier:  Cayman Chemical 292371 MG
Encompass
A mixture of phosphatidylglycerols isolated from egg a metabolic precursor of cardiolipin a constituent of plant animal and bacterial cell membranes species containing polyunsaturated and monounsaturated fatty acyl chains inhibit and promote proliferation of murine keratinocytes respectively is the second largest lipid component of mammalian lung surfactant comprising 10 of the lipids levels are decreased in the lung surfactant of infants with respiratory distress syndrome
Catalog No. 50-293-7191
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.