Learn More
Cayman Chemical 1 3DIpalmItol2ElaIdol Gl 10mg
SDP
Catalog No. 502937036
Encompass_Preferred
A triacylglycerol; has been synthesized as a partially hydrogenated confectionery fat; when used as 10% of the fat component in cocoa butter, it promotes the conversion of low-melting polymorphic forms of cocoa butter-PEP mixtures to high-melting polymorphic forms
Catalog No. 50-293-7036 Supplier Cayman Chemical Supplier No. 2716510 MG
A triacylglycerol; has been synthesized as a partially hydrogenated confectionery fat; when used as 10% of the fat component in cocoa butter, it promotes the conversion of low-melting polymorphic forms of cocoa butter-PEP mixtures to high-melting polymorphic forms