Learn More
Sigma Organic Chemistry 1- 4- 2-Methyl-1H-imida | 1G | MFCD11841267
Supplier: Sigma Organic Chemistry CBR010011G

1- 4- 2-Methyl-1H-imida , 1G
About this Item:
MDL #: MFCD11841267
Molecular Weight: 205.26
UNSPSC Code: 12352200
Chemical Formula: C11H15N3O
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.