Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4,5,8-Tetrachloroanthraquinone 95.0+%, TCI America™
SDP

Catalog No. T330025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T330025G 25 g
T33005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T330025G Supplier TCI America Supplier No. T330025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 81-58-3
Molecular Formula C14H4Cl4O2
Molecular Weight (g/mol) 345.98
MDL Number MFCD00035801
InChI Key DUJPMUKIEFLXRE-UHFFFAOYSA-N
PubChem CID 66484
IUPAC Name 1,4,5,8-tetrachloro-9,10-dihydroanthracene-9,10-dione
SMILES ClC1=CC=C(Cl)C2=C1C(=O)C1=C(Cl)C=CC(Cl)=C1C2=O

Specifications

Melting Point 342°C
Color Yellow
Quantity 25 g
Formula Weight 345.98
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4,5,8-Tetrachloroanthraquinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.