Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane 98.0+%, TCI America™
SDP

Catalog No. T18771G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18771G 1 g
T1877200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T18771G Supplier TCI America Supplier No. T18771G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 41203-22-9
Molecular Formula C14H36N4
Molecular Weight (g/mol) 260.47
MDL Number MFCD00005106
InChI Key HRFJEOWVAGSJNW-UHFFFAOYNA-R
PubChem CID 123502
IUPAC Name 1,4,8,11-tetramethyl-1,4,8,11-tetraazacyclotetradecane-1,4,8,11-tetraium
SMILES C[NH+]1CCC[NH+](C)CC[NH+](C)CCC[NH+](C)CC1

Specifications

Melting Point 40°C
Color White-Yellow
Quantity 1 g
Synonym 1,4,8,11-tetramethyl-1,4,8,11-tetraazacyclotetradecane, tetramethylcyclam, 1,4,8,11-tetraazacyclotetradecane, 1,4,8,11-tetramethyl, acmc-1ankx, 1,4,8,11-tetramethylcyclam, 1,4,8,11-tetraazacyclotetradecane,1,4,8,11-tetramethyl, 1,4,8,11-tetramethyl-1,4,8, 11-tetraazacyclotetradecane, 1r,4s,8r,11s-1,4,8,11-tetramethyl-1,4,8,11-tetraazacyclotetradecane, 1alpha,4beta,8beta,11alpha-tetramethyl-1,4,8,11-tetraazacyclotetradecane
Formula Weight 256.44
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane
RTECSNumber XA5255700
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.