Learn More
Sigma Organic Chemistry 1-4-Aminobutyl -5-bromo 250MG
Supplier: Sigma Organic Chemistry OTV000420250MG

1-4-Aminobutyl -5-bromo , 250mg
About this item:
MDL #: MFCD21090493
Chemical Formula: C9H14BrClN2O
Molecular Weight: 281.58
UNSPSC Code: 12352200
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.