Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Benzodioxane-5-carboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. B386425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B386425G 25 g
B38645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B386425G Supplier TCI America Supplier No. B386425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4442-53-9
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.159
MDL Number MFCD00239415
InChI Key VCLSWKVAHAJSFL-UHFFFAOYSA-N
Synonym 1,4-benzodioxan-5-carboxylic acid, 2,3-dihydrobenzo b 1,4 dioxine-5-carboxylic acid, 2,3-dihydro-1,4-benzodioxin-5-carboxylic acid, 1,4-benzodioxane-5-carboxylic acid, 2,3-dihydro-benzo 1,4 dioxine-5-carboxylic acid, 2,3-ethylenedioxybenzoic acid, 2,3-dihydro-1,4-benzodioxane-5-carboxylic acid, 2,3-dihydrobenzo 1,4 dioxine-5-carboxylic acid, 1,4-benzodioxin-5-carboxylic acid, 2,3-dihydro, 2h,3h-benzo e 1,4-dioxin-5-carboxylic acid
PubChem CID 78184
IUPAC Name 2,3-dihydro-1,4-benzodioxine-5-carboxylic acid
SMILES C1COC2=C(C=CC=C2O1)C(=O)O

Specifications

Melting Point 192°C
Quantity 25 g
Formula Weight 180.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Benzodioxane-5-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.