Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Bis(acryloyloxy)butane (stabilized with MEHQ) 90.0+%, TCI America™
SDP

Catalog No. B2935100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2935100G 100 g
B293525G 25 g
B2935500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B2935100G Supplier TCI America Supplier No. B2935100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1070-70-8
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.22
MDL Number MFCD00014940
InChI Key JHWGFJBTMHEZME-UHFFFAOYSA-N
Synonym 1,4-butanediol diacrylate, butylene diacrylate, butanediol diacrylate, tetramethylene glycol diacrylate, 1,4-butylene glycol diacrylate, tetramethylene acrylate, tetramethylene diacrylate, 1,4-butylene diacrylate, 1,4-butanediyl diacrylate, 1,4-butanediyl bisacrylate
PubChem CID 70613
IUPAC Name 4-(prop-2-enoyloxy)butyl prop-2-enoate
SMILES C=CC(=O)OCCCCOC(=O)C=C

Specifications

Color Yellow
Boiling Point 138°C
Quantity 100 g
UN Number 1760
Formula Weight 198.22
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Bis(acryloyloxy)butane (stabilized with MEHQ)
EINECSNumber (2)-1007&(2)-0958
RTECSNumber UD3130000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.