Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Bis(glycidyloxy)benzene, 97%, Thermo Scientific™

Catalog No. AAB2062803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2062803 1 g
AAB2062806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2062803 Supplier Thermo Scientific Chemicals Supplier No. B2062803

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
is a good general-purpose inhibitor, stabilizer, antioxidant, and intermediate. It is used as a photographic reducer and developer.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2425-01-6
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00186891
InChI Key FSYPIGPPWAJCJG-UHFFFAOYNA-N
Synonym 1,4-bis glycidyloxy benzene, hydroquinone diglycidyl ether, 2-4-oxiran-2-ylmethoxy phenoxymethyl oxirane, hydroquinone 1,4-diglycidyl ether, oxirane, 2,2'-1,4-phenylenebis oxymethylene bis, 2,2'-1,4-phenylenebis oxymethylene dioxirane, 2-4-oxiran-2-ylmethoxy phenoxy methyl oxirane, 1,4-bis oxiran-2-ylmethoxy benzene, acmc-20ap2z
PubChem CID 62443
IUPAC Name 2-[[4-(oxiran-2-ylmethoxy)phenoxy]methyl]oxirane
SMILES C1C(O1)COC2=CC=C(C=C2)OCC3CO3

Specifications

Melting Point 110°C to 112°C
Quantity 1 g
Formula Weight 222.24
Percent Purity 97%
Chemical Name or Material 1,4-Bis(glycidyloxy)benzene
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.