Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Bromo-2-thienyl)ethan-1-one, Tech., Thermo Scientific™

Catalog No. KM10200EA
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
KM10200EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. KM10200EA Supplier Thermo Scientific Chemicals Supplier No. KM10200EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7209-11-2
Molecular Formula C6H5BrOS
Molecular Weight (g/mol) 205.07
MDL Number MFCD01566552
InChI Key HXVLWNKFMNRJED-UHFFFAOYSA-N
Synonym 1-4-bromothiophen-2-yl ethanone, 2-acetyl-4-bromothiophene, 1-4-bromo-2-thienyl ethan-1-one, 1-4-bromo-2-thienyl-1-ethanone, 1-4-bromothiophen-2-yl ethan-1-one, 4-bromo-2-acetylthiophene, 1-4-bromo-2-thienyl ethanone, ethanone,1-4-bromo-2-thienyl, ethanone, 1-4-bromo-2-thienyl, 4-bromo-2-acetyl-thiophene
PubChem CID 2735237

Specifications

Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 205.07
Percent Purity Tech.
Grade Technical
Chemical Name or Material 1-(4-Bromo-2-thienyl)ethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.