Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Bromophenyl)-2-methyl-1-propanone 95.0+%, TCI America™
SDP

Catalog No. B52325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B52325G 5 g
B52321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B52325G Supplier TCI America Supplier No. B52325G

Chemical Identifiers

CAS 49660-93-7
Molecular Formula C10H11BrO
Molecular Weight (g/mol) 227.101
MDL Number MFCD09891214
InChI Key PMFLKZHWZXWOEH-UHFFFAOYSA-N
PubChem CID 15074783
IUPAC Name 1-(4-bromophenyl)-2-methylpropan-1-one
SMILES CC(C)C(=O)C1=CC=C(C=C1)Br

Specifications

Melting Point 18°C
Color Yellow
Boiling Point 87°C
Quantity 5 g
Formula Weight 227.10
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(4-Bromophenyl)-2-methyl-1-propanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.