Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,4-Butane sultone

Catalog No. B85501100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B85501100G 100 g
B8550125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B85501100G Supplier Sigma Aldrich Supplier No. B85501100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,4-Butane sultone

Specifications

CAS 1633-83-6
Melting Point 12°C to 15°C (lit.)
Density 1.331 g/mL (at 25°C (literature))
Boiling Point >165°C (25 mmHg)
Molecular Formula C4H8O3S
Linear Formula C4H8O3S
Refractive Index n20/D 1.464 (literature)
MDL Number MFCD00006584
Quantity 100 g
Synonym 4-Hydroxybutane-1-sulfonic acid delta-sultone
Molecular Weight (g/mol) 136.17
Percent Purity ≥99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.