Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4 Butanediol, MP Biomedicals™

Catalog No. MP230002301 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 4 L
6 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP230002301 1 L
MP230002304 4 L
MP230002316 4 x 4 L
MP230002306 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MP230002301 Supplier MP Biomedicals, Inc Supplier No. 0230002301
 Restricted Item
Add to Cart
Add to Cart

1,4 Butanediol Certified Grade, Clear Oily Liquid

  • 1,4 Butanediol and its derivatives are used in the manufacturing of technical plastics, polyurethanes, solvents, electronic chemicals and elastic fibers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-63-4
Molecular Weight (g/mol) 90.12
MDL Number MFCD00002968
InChI Key WERYXYBDKMZEQL-UHFFFAOYSA-N
Synonym Butane-1,4-diol, 1,4-Butyleneglycol, 1,4-Dihdyroxybutane, Tetramethylene 1,4-diol
IUPAC Name butane-1,4-diol
SMILES OCCCCO

Specifications

Melting Point 16°C (literature)
Color ≤10 (APHA)
Density 1.017 g/mL at 25°C (Lit.)
Boiling Point 230°C (Literature)
Flash Point 110°C / 273.2°F (Lit.)
Molecular Formula C4H10O2
Refractive Index n20/D 1.445 (Lit.)
Quantity 1 L
Vapor Density 3.1g/L (vs air)(Lit.)
Vapor Pressure < 1 hPa at 20°C (Lit.)
Grade Certified
Physical Form Oily Liquid
Show More Show Less
RTECSNumber EK0525000
storageNote1 Store at Room Temperature (15-30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.