Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4 Butanediol, MP Biomedicals™

Catalog No. MP230002306 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 4 L
6 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP230002306 6 x 1 L
MP230002301 1 L
MP230002304 4 L
MP230002316 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MP230002306 Supplier MP Biomedicals, Inc Supplier No. 0230002306

1,4 Butanediol Certified Grade, Clear Oily Liquid

  • 1,4 Butanediol and its derivatives are used in the manufacturing of technical plastics, polyurethanes, solvents, electronic chemicals and elastic fibers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Weight (g/mol) 90.12
MDL Number MFCD00002968
Synonym Butane-1,4-diol, 1,4-Butyleneglycol, 1,4-Dihdyroxybutane, Tetramethylene 1,4-diol

Specifications

Melting Point 16°C (literature)
Color ≤10 (APHA)
Density 1.017g/mL at 25°C (Lit.)
Boiling Point 230°C (Literature)
Flash Point 110°C / 273.2°F (Lit.)
Molecular Formula HO(CH2)4OH
Refractive Index n20/D 1.445 (Lit.)
Quantity 6 x 1 L
Beilstein 1633445
Vapor Density 3.1g/L (vs air)(Lit.)
Vapor Pressure < 1 hPa at 20°C (Lit.)
Grade Certified
Physical Form Oily Liquid
Assay ≥99.0%
Show More Show Less
RTECSNumber EK0525000
storageNote1 Store at Room Temperature (15-30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.