Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Butanedithiol 95.0+%, TCI America™
SDP

Catalog No. B082925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B082925ML 25 mL
B08295ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B082925ML Supplier TCI America Supplier No. B082925ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1191-08-8
Molecular Formula C4H10S2
Molecular Weight (g/mol) 122.24
MDL Number MFCD00004906
InChI Key SMTOKHQOVJRXLK-UHFFFAOYSA-N
Synonym 1,4-butanedithiol, 1,4-dimercaptobutane, tetramethylenedithiol, unii-7jd227iumc, tetramethylene dimercaptan, 7jd227iumc, 1,4-butane dithiol, 1,4-dibutyl mercaptan, acmc-1bs9f
PubChem CID 79148
IUPAC Name butane-1,4-dithiol
SMILES SCCCCS

Specifications

Color Yellow
Boiling Point 195°C
Quantity 25 mL
Formula Weight 122.24
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1,4-Butanedithiol
EINECSNumber (2)-2448
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.