Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Butanesultone 99.0+%, TCI America™
SDP

Catalog No. B0136250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0136250G 250 g
B013625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0136250G Supplier TCI America Supplier No. B0136250G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1633-83-6
Molecular Formula C4H8O3S
Molecular Weight (g/mol) 136.17
MDL Number MFCD00006584
InChI Key MHYFEEDKONKGEB-UHFFFAOYSA-N
Synonym 1,4-butane sultone, 1,4-butanesultone, butane sultone, butanesultone, 1,2-oxathiane, 2,2-dioxide, butanesulfone, 1,2-oxathiane 2,2-dioxide, 1,4-butylene sulfone, delta-valerosultone, 1,4-butanesulfone
PubChem CID 15411
IUPAC Name 1,2λ⁶-oxathiane-2,2-dione
SMILES O=S1(=O)CCCCO1

Specifications

Color Colorless
Boiling Point 146°C
Quantity 250 g
Formula Weight 136.17
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Butanesultone
EINECSNumber (5)-0531
RTECSNumber RP4300000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.