Learn More
AdipoGen 1-4-Carboxybutyl-3 7-dmx
SDP
Supplier:  AdipoGen CDXC0312M010
Encompass_Preferred
Chemical. CAS 38975-44-9. Formula C12H16N4O4. MW 280.28. Synthetic Pentoxifylline PTX metabolite. Can be used as reference material. Showed platelet aggregation in whole blood in vitro. Shown to be more effective than PTX in protecting WC/1 murine fibrosarcoma cells from TNF-alpha cytotoxicity and in inhibiting neutrophil superoxide anion production, degranulation lactoferrin release and surface expression of the beta-2 integrin CD11b/CD18.
Catalog No. 50-256-1385
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.