Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-(4-chlorophenyl)ethanol | 3391-10-4 | MFCD00041038 | 99.2% | 156.61 g/mol | C8H9ClO | 25 G
SDP

Supplier:  Medchemexpress LLC 25GHYW074486

Encompass_Preferred

1-(4-chlorophenyl)ethanol is an organic biochemical reagent used as a reference standard and synthetic intermediate in assay development and research. It is supplied as a liquid, identity-confirmed by 1H NMR and mass spectrometry, and characterized by high HPLC purity.

  • Colorless to light yellow liquid appearance.
  • High HPLC purity (COA batch result 99.24%, reported as 99.2%).
  • Molecular formula C8H9ClO; molecular weight 156.61 g/mol.
  • Identity confirmed by 1H NMR and MS.
  • Available in multiple pack sizes, including 25 G.
  • Storage: room temperature for up to 3 years; in solvent: -80°C (2 years), -20°C (1 year).

Catalog No. 50-002-09551


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.