Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Cyanophenyl)guanidine 99.0+%, TCI America™
SDP

Catalog No. C33871G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C33871G 1 g
C33875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C33871G Supplier TCI America Supplier No. C33871G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5637-42-3
Molecular Formula C8H8N4
Molecular Weight (g/mol) 160.18
MDL Number MFCD16251323
InChI Key BTDGLZSKNFJBER-UHFFFAOYSA-N
Synonym 1-4-cyanophenyl guanidine, n-4-cyanophenyl guanidine, 2-4-cyanophenyl guanidine, 4-cyanophenyl guanidine, 4-cyanophenylguanidine
PubChem CID 17747914
IUPAC Name 2-(4-cyanophenyl)guanidine
SMILES C1=CC(=CC=C1C#N)N=C(N)N

Specifications

Melting Point 212°C
Color White-Yellow
Quantity 1 g
Formula Weight 160.18
Percent Purity ≥99.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Cyanophenyl)guanidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.