Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Cyclohexadiene, 97% stab. with 0.1% BHT

Catalog No. AAL0733714
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0733714 25 g
AAL0733706 5 g
AAL0733722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0733714 Supplier Thermo Scientific Chemicals Supplier No. L0733714
Add to Cart

CAS: 628-41-1 | C6H8 | 80.13 g/mol

1,4-Cyclohexadiene is used to study the formation of parent ion from heavy fragmentation on irradiation with a high-intensity laser pulse. It is also used as a very effective hydrogen donor for catalytic hydrogenation reaction. It forms benzene at elevated temperatures in the presence of a ruthenium(II)-triphenylphosphine catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Cyclohexadiene is used to study the formation of parent ion from heavy fragmentation on irradiation with a high-intensity laser pulse. It is also used as a very effective hydrogen donor for catalytic hydrogenation reaction. It forms benzene at elevated temperatures in the presence of a ruthenium(II)-triphenylphosphine catalyst.

Solubility
Miscible with organic solvents like diethyl ether, tetrahydrofuran and toluene.Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-41-1
Molecular Formula C6H8
Molecular Weight (g/mol) 80.13
MDL Number MFCD00001535
InChI Key UVJHQYIOXKWHFD-UHFFFAOYSA-N
Synonym 1,4-cyclohexadiene, 1,4-dihydrobenzene, 1,4-cyclohexanediene, unii-0f8z5909qz, 1,4 cyclohexadiene, acmc-209snj, ksc354m7f, 1,4-cyclohexadiene, stabilized 5g, 1,4-cyclohexadiene, purum gc
PubChem CID 12343
ChEBI CHEBI:37611
IUPAC Name cyclohexa-1,4-diene
SMILES C1C=CCC=C1

Specifications

Melting Point -49°C
Density 0.847
Boiling Point 88°C to 90°C
Flash Point −6°C (21°F)
Refractive Index 1.472
Quantity 25 g
UN Number UN3295
Beilstein 1900733
Solubility Information Miscible with organic solvents like diethyl ether,tetrahydrofuran and toluene.Immiscible with water.
Formula Weight 80.13
Percent Purity 97%
Chemical Name or Material 1,4-Cyclohexadiene, stabilized with 0.1% BHT
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225-H340-H350-H304
Highly flammable liquid and vapor.
May cause genetic defects.
May cause cancer.
May be fatal if swallowed and enters airways.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 211-043-1
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.