Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Cyclohexanediamine (cis- and trans- mixture) 97.0+%, TCI America™
SDP

Catalog No. C081425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C081425ML 25 mL
C08145ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C081425ML Supplier TCI America Supplier No. C081425ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3114-70-3
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.19
MDL Number MFCD00035862,MFCD00075174,MFCD08276323
InChI Key VKIRRGRTJUUZHS-UHFFFAOYSA-N
Synonym 1,4-Diaminocyclohexane
PubChem CID 18374
IUPAC Name cyclohexane-1,4-diamine
SMILES NC1CCC(N)CC1

Specifications

Color White-Yellow
Boiling Point 197°C
Quantity 25 mL
UN Number 3259
Formula Weight 114.19
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 1,4-Cyclohexanediamine (cis- and trans- mixture)
RTECSNumber GU8750500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.