Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Cyclohexanedione Monoethyleneketal 98.0+%, TCI America™
SDP

Catalog No. C129225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C129225G 25 g
C12925G 5 g
C1292100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C129225G Supplier TCI America Supplier No. C129225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4746-97-8
Molecular Formula C8H12O3
Molecular Weight (g/mol) 156.181
MDL Number MFCD00010214
InChI Key VKRKCBWIVLSRBJ-UHFFFAOYSA-N
Synonym 1,4-dioxaspiro 4.5 decan-8-one, 1,4-cyclohexanedione monoethylene acetal, 1,4-cyclohexanedione monoethylene ketal, 1,4-cyclohexanedione monoethyleneacetal, 1,4-dioxa-spiro 4.5 decan-8-one, 1,4-cyclohexanedione monoethyleneketal, cyclohexane-1,4-dione monoethylene ketal, 1,4-dioxaspiro 4.5 decane-8-one
PubChem CID 567415
IUPAC Name 1,4-dioxaspiro[4.5]decan-8-one
SMILES C1CC2(CCC1=O)OCCO2

Specifications

Melting Point 73°C
Quantity 25 g
Formula Weight 156.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Cyclohexanedione Monoethyleneketal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.