Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-1,4-Di-O-tosyl-2,3-O-isopropylidene-D-threitol 97.0+%, TCI America™
SDP

Catalog No. D16221G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D16221G Supplier TCI America Supplier No. D16221G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 51064-65-4
Molecular Formula C21H26O8S2
Molecular Weight (g/mol) 470.551
MDL Number MFCD00063235
InChI Key KPFDKWNWYAXRNJ-WOJBJXKFSA-N
Synonym +-1,4-di-o-tosyl-2,3-o-isopropylidene-d-threitol, 1,4-di-o-tosyl-2,3-o-isopropylidene-d-threitol, 2r,3r-1,4-di-o-tosyl-2,3-o-isopropylidene-d-threitol, 4r-trans-2,2-dimethyl-1,3-dioxolane-4,5-dimethyl bis toluene-p-sulphonate, 4r,5r-2,2-dimethyl-1,3-dioxolane-4,5-diyl bis methylene bis 4-methylbenzenesulfonate, --1,4-di-o-tosyl-2,3-o-isopropylidenethreitol, +-2,3-o-isopropylidene-1,4-di-o-tosyl-d-threitol, 4r,5r-2,2-dimethyl-5-4-methylbenzenesulfonyl oxy methyl-1,3-dioxolan-4-yl methyl 4-methylbenzenesulfonate, pubchem6770, r,r-+-1,4-di-o-tosyl-2,3-o-isopropylidene-d-threitol
PubChem CID 97446
IUPAC Name [(4R,5R)-2,2-dimethyl-5-[(4-methylphenyl)sulfonyloxymethyl]-1,3-dioxolan-4-yl]methyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCC2C(OC(O2)(C)C)COS(=O)(=O)C3=CC=C(C=C3)C

Specifications

Melting Point 90°C
Color White-Yellow
Quantity 1 g
Formula Weight 470.55
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (+)-1,4-Di-O-tosyl-2,3-O-isopropylidene-D-threitol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.