Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Di-tert-butylbenzene 99.0+%, TCI America™
SDP

Catalog No. D211125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D211125G 25 g
D21115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D211125G Supplier TCI America Supplier No. D211125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1012-72-2
Molecular Formula C14H22
Molecular Weight (g/mol) 190.33
MDL Number MFCD00008836
InChI Key OOWNNCMFKFBNOF-UHFFFAOYSA-N
Synonym 1,4-di-tert-butylbenzene, p-di-tert-butylbenzene, benzene, p-di-tert-butyl, benzene, 1,4-bis 1,1-dimethylethyl, unii-55ptx4kh71, bis 1,1-dimethylethyl benzene, benzene, bis 1,1-dimethylethyl, 1,4-di-t-butylbenzene, acmc-2097up, 1,4-bis tert-butyl benzene
PubChem CID 13895
IUPAC Name 1,4-ditert-butylbenzene
SMILES CC(C)(C)C1=CC=C(C=C1)C(C)(C)C

Specifications

Melting Point 78°C
Color White
Boiling Point 236°C
Quantity 25 g
Formula Weight 190.33
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Di-tert-butylbenzene
EINECSNumber (3)-0025
RTECSNumber CY8444000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.