Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Diazabicyclo[2.2.2]octane Dihydrochloride 98.0+%, TCI America™
SDP

Catalog No. D52511G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D52511G 1 g
D52515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D52511G Supplier TCI America Supplier No. D52511G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 49563-87-3
Molecular Formula C6H14Cl2N2
Molecular Weight (g/mol) 185.092
InChI Key FXNCMDQKBKIDGU-UHFFFAOYSA-N
PubChem CID 12254369
IUPAC Name 1,4-diazabicyclo[2.2.2]octane;dihydrochloride
SMILES C1CN2CCN1CC2.Cl.Cl

Specifications

Melting Point 320°C
Color White
Quantity 1 g
Formula Weight 185.09
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Diazabicyclo[2.2.2]octane Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.