Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Diazabicyclo-[2.2.2]-Octane, MP Biomedicals

Catalog No. MP219513725 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219513725 25 g
MP219513780 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP219513725 Supplier MP Biomedicals, Inc Supplier No. 0219513725

  • 1,4-Diazabicyclo-(2,2,2)-octane is an anti-fade reagent that scavenges free-radicals produced by the excitation of fluorochromes
  • Added to the mounting medium in fluorescence microscopy to retard photo bleaching of fluorescein and other fluorescent dyes
  • It is also an oxidation and polymerization catalyst

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 280-57-9
Molecular Formula C6H12N2
Molecular Weight (g/mol) 112.176
MDL Number MFCD00006689
InChI Key IMNIMPAHZVJRPE-UHFFFAOYSA-N
Synonym 1,4-diazabicyclo 2.2.2 octane, triethylenediamine, dabco, dabco 33lv, 1,4-ethylenepiperazine, dabco crystal, teda, texacat td 100, dabco s-25
PubChem CID 9237
IUPAC Name 1,4-diazabicyclo[2.2.2]octane
SMILES C1CN2CCN1CC2

Specifications

Melting Point 150°C to 160°C
Color Yellow
Boiling Point 174°C (literature)
Linear Formula C6H12N2
Quantity 25 g
Solubility Information Soluble in water, acetone, benzene, ethanol, methyl ethyl, ketone.
Formula Weight 112.18
Physical Form Crystalline Powder
Chemical Name or Material 1,4-DIAZABICYCLO-[2.2.2]-OCTANE
Recommended Storage +4°C, desiccate, store under nitrogen.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.