Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dibromo-2,5-diiodobenzene 96.0+%, TCI America™
SDP

Catalog No. D390425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D390425G 25 g
D39045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D390425G Supplier TCI America Supplier No. D390425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 63262-06-6
Molecular Formula C6H2Br2I2
Molecular Weight (g/mol) 487.699
MDL Number MFCD00082898
InChI Key IVKPEQAIHJWGGT-UHFFFAOYSA-N
PubChem CID 11123771
IUPAC Name 1,4-dibromo-2,5-diiodobenzene
SMILES C1=C(C(=CC(=C1I)Br)I)Br

Specifications

Melting Point 167°C
Color White
Quantity 25 g
Formula Weight 487.70
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Dibromo-2,5-diiodobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.