Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Diformylpiperazine, 98+%, Thermo Scientific™

Catalog No. AAA1809722
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1809722 100 g
AAA1809714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1809722 Supplier Thermo Scientific Chemicals Supplier No. A1809722

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4164-39-0
Molecular Formula C6H10N2O2
Molecular Weight (g/mol) 142.16
MDL Number MFCD00006153
InChI Key CBLGQEBXWDKYDI-UHFFFAOYSA-N
Synonym 1,4-diformylpiperazine, 1,4-piperazinedicarboxaldehyde, pubchem15032, acmc-1ago2, 1,4-piperazine dialdehyde, piperazine-1,4-dicarboxaldehyde, 1,4 piperazinedicarboxaldehyde
PubChem CID 77821
IUPAC Name piperazine-1,4-dicarbaldehyde
SMILES O=CN1CCN(CC1)C=O

Specifications

Melting Point 126°C to 129°C
Boiling Point 154°C (0.25 mmHg)
Odor Odorless
Quantity 100 g
Beilstein 121175
Sensitivity Air Sensitive
Formula Weight 142.16
Percent Purity ≥98%
Chemical Name or Material 1,4-Diformylpiperazine
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
EINECSNumber 224-011-7
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.