CAS RN 4164-39-0
1,4-Diformylpiperazine, TCI America™
CAS: 4164-39-0 Molecular Formula: C6H10N2O2 Molecular Weight (g/mol): 142.16 MDL Number: MFCD00006153 InChI Key: CBLGQEBXWDKYDI-UHFFFAOYSA-N Synonym: 1,4-diformylpiperazine,1,4-piperazinedicarboxaldehyde,pubchem15032,acmc-1ago2,1,4-piperazine dialdehyde,piperazine-1,4-dicarboxaldehyde,1,4 piperazinedicarboxaldehyde PubChem CID: 77821 IUPAC Name: piperazine-1,4-dicarbaldehyde SMILES: O=CN1CCN(CC1)C=O
MP Biomedicals, Inc 1,4-Piperazine Dialdehyde, MP Biomedicals
CAS: 4164-39-0 Molecular Formula: C6H10N2O2 Molecular Weight (g/mol): 142.16 MDL Number: MFCD00006153 InChI Key: CBLGQEBXWDKYDI-UHFFFAOYSA-N Synonym: 1,4-diformylpiperazine,1,4-piperazinedicarboxaldehyde,pubchem15032,acmc-1ago2,1,4-piperazine dialdehyde,piperazine-1,4-dicarboxaldehyde,1,4 piperazinedicarboxaldehyde PubChem CID: 77821 IUPAC Name: piperazine-1,4-dicarbaldehyde SMILES: O=CN1CCN(CC1)C=O
1,4-Diformylpiperazine, 98+%, Thermo Scientific™
CAS: 4164-39-0 Molecular Formula: C6H10N2O2 Molecular Weight (g/mol): 142.16 MDL Number: MFCD00006153 InChI Key: CBLGQEBXWDKYDI-UHFFFAOYSA-N Synonym: 1,4-diformylpiperazine,1,4-piperazinedicarboxaldehyde,pubchem15032,acmc-1ago2,1,4-piperazine dialdehyde,piperazine-1,4-dicarboxaldehyde,1,4 piperazinedicarboxaldehyde PubChem CID: 77821 IUPAC Name: piperazine-1,4-dicarbaldehyde SMILES: O=CN1CCN(CC1)C=O