Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Diformylpiperazine, TCI America™
SDP

Catalog No. D257825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D257825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D257825G Supplier TCI America Supplier No. D257825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4164-39-0
Molecular Formula C6H10N2O2
Molecular Weight (g/mol) 142.16
MDL Number MFCD00006153
InChI Key CBLGQEBXWDKYDI-UHFFFAOYSA-N
Synonym 1,4-diformylpiperazine, 1,4-piperazinedicarboxaldehyde, pubchem15032, acmc-1ago2, 1,4-piperazine dialdehyde, piperazine-1,4-dicarboxaldehyde, 1,4 piperazinedicarboxaldehyde
PubChem CID 77821
IUPAC Name piperazine-1,4-dicarbaldehyde
SMILES O=CN1CCN(CC1)C=O

Specifications

Melting Point 127°C
Color White
Quantity 25 g
Formula Weight 142.16
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Diformylpiperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.