Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dimethoxy-2-butyne 98.0+%, TCI America™
SDP

Catalog No. D49511ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49511ML 1 mL
D49515ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49511ML Supplier TCI America Supplier No. D49511ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16356-02-8
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00048069
InChI Key YFUQYYGBJJCAPR-UHFFFAOYSA-N
PubChem CID 542354
IUPAC Name 1,4-dimethoxybut-2-yne
SMILES COCC#CCOC

Specifications

Melting Point -39°C
Color Yellow
Boiling Point 157°C
Quantity 1 mL
UN Number 3271
Formula Weight 114.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Dimethoxy-2-butyne
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.