Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dimethyl-1,2,4-triazolium Iodide 98.0+%, TCI America™
SDP

Catalog No. D39621G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39621G 1 g
D39625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39621G Supplier TCI America Supplier No. D39621G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 120317-69-3
Molecular Formula C4H8IN3
Molecular Weight (g/mol) 225.033
InChI Key IMFBMMNQDMTHGG-UHFFFAOYSA-M
PubChem CID 10987916
IUPAC Name 1,4-dimethyl-1,2,4-triazol-4-ium;iodide
SMILES CN1C=[N+](C=N1)C.[I-]

Specifications

Melting Point 118°C
Color Yellow
Quantity 1 g
Formula Weight 225.03
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Dimethyl-1,2,4-triazolium Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.