Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1,4-dimethylpiperazin-2-yl)methylamine, Thermo Scientific™

Catalog No. CC13813CB
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC13813CB 250 mg Amber Glass Bottle
CC13813DA 1 g Amber Glass Bottle
CC13813DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC13813CB Supplier Thermo Scientific Chemicals Supplier No. CC13813CB
Add to Cart

Chemical Identifiers

CAS 131922-07-1
Molecular Formula C7H17N3
Molecular Weight (g/mol) 143.234
InChI Key DKFODFYKOIZTMO-UHFFFAOYSA-N
Synonym 1,4-dimethylpiperazin-2-yl methanamine, 1,4-dimethylpiperazin-2-yl methylamine, 1,4-dimethylpiperazin-2-yl methyl amine, 1-1,4-dimethyl-2-piperazinyl methanamine, 1-1,4-dimethylpiperazin-2-yl methanamine
PubChem CID 10464433
IUPAC Name (1,4-dimethylpiperazin-2-yl)methanamine
SMILES CN1CCN(C(C1)CN)C

Specifications

Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 143.23
Percent Purity 97%
Chemical Name or Material (1, 4-dimethylpiperazin-2-yl)methylamine
missing translation for 'healthHazard1' GHS Signal Word: Danger
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.