Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dioxane, Reagent Grade, ≥99%, Solstice

Catalog No. 60047943 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
20 L
Packaging:
Stainless Steel Can
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-943 20 L Stainless Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-047-943 Supplier Solstice Supplier No. D20186320L

missing translation for 'validMainframeMsg'

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-91-1
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00006571
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
Synonym dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
PubChem CID 31275
ChEBI CHEBI:47032
IUPAC Name 1,4-dioxane
SMILES C1COCCO1

Specifications

pH 6 to 8 (at 500.00g/L 20°C)
Melting Point 12°C
Color Colorless
Density 1.034g/mL at 25°C
Quantity 20 L
Explosion Limit 0.22
Flash Point 12°C (53.6°F)
Ignition Point 356°F
Odor Aromatic
Packaging Stainless Steel Can
Physical Form Liquid
Vapor Pressure 27mmHg (20°C), 40mmHg (25°C)
UN Number UN1165
Beilstein 102551
Boiling Point 100°C to 102°C (1013 hPa)
Solubility Information Fully miscible in water
Formula Weight 88.11g/mol
Percent Purity ≥99%
Chemical Name or Material 1,4-Dioxane
Show More Show Less
Hazard Statement P210-P261-P281-P305 + P351 + P338
DOTInformation class 3 - PG 2 - Dioxane
RTECSNumber 204-661-8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.