Learn More
Sigma Aldrich Fine Chemicals Biosciences 1,4-Dioxane, 123-91-1, MFCD00006571, 100 mL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 296309100ML
Encompass
Empirical Formula (Hill Notation): C4H8O2, Molecular Weight: 88.11, anhydrous, 99.8%, Synonym: Diethylene oxide, Dioxane. 1,4-Dioxane may undergo nickel-catalyzed oxidative arylation with arylboronic acids to form α-arylated ethers.
Catalog No. NC0880964
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.