Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dioxane, for HPLC, GC and spectrophotometry, >99.9%, Solstice

Catalog No. 6000081 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
Packaging:
Glass Bottle
Poly-coated Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-000-81 4 x 4 L Glass Bottle
60-000-82 4 x 4 L Poly-coated Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-000-81 Supplier Solstice Supplier No. 0874

Chemical Identifiers

CAS 123-91-1
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
Synonym dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
PubChem CID 31275
ChEBI CHEBI:47032
IUPAC Name 1,4-dioxane
SMILES C1COCCO1

Specifications

Absorbance 0.3 AU max. at 250nm (UV), 0.02 AU max. at 300nm (UV), 0.005 AU max. at 350nm (UV), 0.005 AU max. at 400nm (UV)
CAS Max % 100%
Density 1.0280 g/cm3
Quantity 4 x 4 L
Flash Point 12°C
Packaging Glass Bottle
Water 0.05% max.
UN Number UN1165
Beilstein 102551
Boiling Point 101.32°C
Solubility Information Miscible in Water at 20°C
Formula Weight 88.11
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material 1,4-Dioxane, B&J Brand™, for HPLC, GC and spectrophotometry, >99.9%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Dioxane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.