Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dioxane-d{8}, 99% (Isotopic)

Catalog No. aa3651603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3651603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3651603 Supplier Thermo Scientific Chemicals Supplier No. 03651603
Add to Cart

CAS: 17647-74-4 | C4H8O2 | 96.155 g/mol

1,4-Dioxane-d{8} is used as a solvent in nuclear magnetic resonance (NMR) spectroscopy and acts as a stabilizer for 1,1,1-trichloroethane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17647-74-4
Molecular Formula C4H8O2
Molecular Weight (g/mol) 96.155
MDL Number MFCD00044239
InChI Key RYHBNJHYFVUHQT-SVYQBANQSA-N
Synonym 1,4-dioxane-d8, 2h8-1,4-dioxane, 1,4-dioxane-d8, 99 atom % d, 2 h?-1,4-dioxane, 1,4-dioxane-2lh8, 1,4-dioxane d8, 1,4-dioxane-d8, ≥99 atom % d, 1,4-dioxane-d isotopic 1g, 1,4-dioxane-d8 >99.0 atom % d, 1 4-dioxane-d8 99 atom % d
PubChem CID 87209
IUPAC Name 2,2,3,3,5,5,6,6-octadeuterio-1,4-dioxane
SMILES C1COCCO1

Specifications

Physical Form Liquid
Quantity 1 g
Density 1.13 g/mL
Sensitivity Hygroscopic
Melting Point 12°C
Boiling Point 101°C
UN Number UN1165
Solubility Information Miscible with water.
Formula Weight 96.16
Percent Purity 99%
Chemical Name or Material 1,4-Dioxane-d8, Isotopic
Show More Show Less
missing translation for 'healthHazard1' H225-H319-H335
missing translation for 'healthHazard2' GHS H Statement
H225-H351-H319-H335
Highly flammable liquid and vapor.
Suspected of causing cancer.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DIOXANE
EINECSNumber 241-628-7
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.