Learn More
1,4-Dioxane, HPLC Grade, 99% min, packaged under Argon in resealable ChemSeal™ bottles
Description
1,4-Dioxane HPLC Grade is used as a mobile phase in high performance liquid chromatography and liquid chromatography coupled with mass spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 11.8°C |
| Density | 1.034 |
| Quantity | 250 mL |
| Flash Point | 12°C (54°F) |
| Physical Form | Liquid |
| Refractive Index | 1.422 |
| Assay Percent Range | 99% min |
| Beilstein | 102551 |
| Boiling Point | 100°C to 102°C |
| Solubility Information | Miscible with water,most organic solvents,aromatic hydrocarbons and oils. |
| Show More |
Safety and Handling
| Hazard Category | H225-H319-H335-H336-H350-H373 |
| Precautionary Statement | P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280i-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P370+P378q-P501c |
| DOTInformation | Hazard Class: 3; Packaging Group: II |
| EINECSNumber | 204-661-8 |
| RTECSNumber | JG8225000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.