Learn More
1,4-Dioxane in Water, WS Proficiency Testing Material, MilliporeSigma™ Supelco™
Catalog No. 111017308 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
2 mL
Catalog No. Quantity
11-101-7308 2 mL
1 options
Catalog No. 11-101-7308 Supplier MilliporeSigma Supelco Supplier No. PE15192ML
This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 2 mL
Concentration or Composition (by Analyte or Components) 1,4-Dioxane (1,4- Diethyleneoxide) 1-20 μg/L