Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Diphenoxybenzene 98.0+%, TCI America™
SDP

Catalog No. D201725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D201725G 25 g
D20175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D201725G Supplier TCI America Supplier No. D201725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3061-36-7
Molecular Formula C18H14O2
Molecular Weight (g/mol) 262.308
MDL Number MFCD00038368
InChI Key UVGPELGZPWDPFP-UHFFFAOYSA-N
Synonym benzene, 1,4-diphenoxy, p-diphenoxybenzene, hydroquinone diphenyl ether, p-phenoxyphenoxybenzene, benzene, p-diphenoxy, 4-phenoxydiphenyl oxide, 1,1'-1,4-phenylenebis oxy dibenzene, 1,4-diphenoxy-benzene, acmc-1cqa0
PubChem CID 520487
ChEBI CHEBI:39271
IUPAC Name 1,4-diphenoxybenzene
SMILES C1=CC=C(C=C1)OC2=CC=C(C=C2)OC3=CC=CC=C3

Specifications

Melting Point 76°C
Color White-Yellow
Boiling Point 182°C
Quantity 25 g
Formula Weight 262.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Diphenoxybenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.