Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Fluoro-2-nitrophenyl)-4-methylpiperazine, 97%, Thermo Scientific™

Catalog No. AAH6319803
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6319803 1 g
AAH63198MD 250 mg
AAH6319806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6319803 Supplier Thermo Scientific Chemicals Supplier No. H6319803

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 432531-43-6
Molecular Formula C11H14FN3O2
Molecular Weight (g/mol) 239.25
MDL Number MFCD03144445
InChI Key SVXUJOMAWXJSIK-UHFFFAOYSA-N
Synonym 1-4-fluoro-2-nitrophenyl-4-methylpiperazine, cambridge id 6838238
PubChem CID 2915664
IUPAC Name 1-(4-fluoro-2-nitrophenyl)-4-methylpiperazine
SMILES CN1CCN(CC1)C2=C(C=C(C=C2)F)[N+](=O)[O-]

Specifications

Quantity 1 g
Formula Weight 239.25
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(4-Fluoro-2-nitrophenyl)-4-methylpiperazine
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.