Learn More
Sigma Organic Chemistry 1- 4-Fluorophenyl -1 4- | 1G | MFCD11841252
Supplier: Sigma Organic Chemistry CBR006011G

1- 4-Fluorophenyl -1 4- , 1G
About this Item:
MDL #: MFCD11841252
Molecular Weight: 254.3
UNSPSC Code: 12352200
Chemical Formula: C13H19FN2O2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.