Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Fluorophenyl)-1-methylethylamine, 97%

Catalog No. AC431060010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC431060010 1 g Glass bottle
AC431060050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC431060010 Supplier Thermo Scientific Chemicals Supplier No. 431060010
Add to Cart

CAS: 17797-10-3 | C9H12FN | 153.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17797-10-3
Molecular Formula C9H12FN
Molecular Weight (g/mol) 153.20
MDL Number MFCD07777176
InChI Key NYFSRXOWJXAHOO-UHFFFAOYSA-N
Synonym 2-4-fluorophenyl propan-2-amine, 1-4-fluorophenyl-1-methylethylamine, 1-4-fluorophenyl-1-methyl-ethylamine, 1-4-fluoro-phenyl-1-methyl-ethylamine, 2-4-fluorophenyl prop-2-ylamine, 1-methyl-1-4-fluorophenyl ethylamine, 4-fluorocumylamine, pubchem16914, 4-fluoro-alpha,alpha-dimethylbenzylamine, alpha,alpha-dimethyl-4-fluorobenzylamine
PubChem CID 13025665
IUPAC Name 2-(4-fluorophenyl)propan-2-amine
SMILES CC(C)(N)C1=CC=C(F)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Boiling Point 40.0°C (7.0 Mbar)
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 153.2
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-(4-Fluorophenyl)-1-methylethylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.