Learn More
Sigma Organic Chemistry 1- 4-Fluorophenyl -1H-p | 1G | MFCD08446364
Supplier: Sigma Organic Chemistry JRD02731G

1-(4-Fluorophenyl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine, 1G
About This Item:
Linear Formula: C11H8FN5
MDL Number: MFCD08446364
UNSPSC Code: 12352200
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.