Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Iodobenzyl)pyrrolidine, ≥97%, Thermo Scientific™

Catalog No. CC55611DE
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC55611DE 5 g
CC55611DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC55611DE Supplier Thermo Scientific Chemicals Supplier No. CC55611DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 858676-60-5
Molecular Formula C11H14IN
Molecular Weight (g/mol) 287.144
MDL Number MFCD07775752
InChI Key RDHZJYCRHTZBIX-UHFFFAOYSA-N
Synonym 1-4-iodobenzyl pyrrolidine, 1-4-iodophenyl methyl pyrrolidine, 1-4-iodo-benzyl-pyrrolidine, pyrrolidine,1-4-iodophenyl methyl
PubChem CID 18525867
IUPAC Name 1-[(4-iodophenyl)methyl]pyrrolidine
SMILES C1CCN(C1)CC2=CC=C(C=C2)I

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 287.14
Percent Purity 97%
Chemical Name or Material 1-(4-Iodobenzyl)pyrrolidine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.